1,3-Dichloro-2,4-difluoro-5-nitrobenzene ≥95%

Supplier: Matrix Scientific

076528-1G 076528-5G 076528-10G
102546-682EA 355.38 USD
102546-682 102546-684 102546-686
1,3-Dichloro-2,4-difluoro-5-nitrobenzene ≥95%
1,3-Dichloro-2,4-difluoro-5-nitrobenzene

Hazard: Irritant

Formula: C₆HCl₂F₂NO₂
CAS Number: 15952-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR