(4-Cyanophenyl)acetic acid methyl ester ≥95%

Supplier: Matrix Scientific

076527-1G 076527-5G 076527-10G
102546-676EA 640 USD
102546-676 102546-678 102546-680
(4-Cyanophenyl)acetic acid methyl ester ≥95%
(4-Cyanophenyl)acetic acid methyl ester

Hazard: Irritant

Formula: C₁₀H₉NO₂
MDL Number: MFCD02181140
CAS Number: 52798-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR