2-Amino-2-cyclohexylacetic acid ≥95%

Supplier: Matrix Scientific

076524-1G 076524-5G
102546-660EA 230.77 USD
102546-660 102546-662
2-Amino-2-cyclohexylacetic acid ≥95%
2-Amino-2-cyclohexylacetic acid

Hazard: Irritant

Formula: C₈H₁₅NO₂
Boiling Pt: 292 °C
Melting Pt: 256 °C
Density: 1.12g/cc
Flash Pt: 130 °C
MDL Number: MFCD05662746
CAS Number: 5664-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR