(3-Chloropyrazin-2-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

076523-1G 076523-5G
102546-654EA 246.15 USD
102546-654 102546-656
(3-Chloropyrazin-2-yl)methanamine hydrochloride ≥95%
(3-Chloropyrazin-2-yl)methanamine hydrochloride

Hazard: Irritant/Store in Freezer

Formula: C₅H₇Cl₂N₃
MW: 180.0352 g/mol
MDL Number: MFCD09910171
CAS Number: 939412-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR