1,2-Diamino-3-chlorobenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chlorobenzene-1,2-diamine
076522-1G
076522-5G
102546-648EA
84.62
USD
102546-648
102546-650
1,2-Diamino-3-chlorobenzene ≥95%
1,2-Diamino-3-chlorobenzene
Hazard: Irritant/Store in Freezer
Formula:
C₆H₇ClN₂ MW: 142.59 g/mol Melting Pt: 27…29 °C |
MDL Number:
MFCD09881742 CAS Number: 21745-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...