6-Bromoisoindolin-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2,3-dihydro-1H-isoindol-1-one

076521-1G 076521-5G
102546-644EA 41.54 USD
102546-644 102546-646
6-Bromoisoindolin-1-one ≥95%
6-Bromoisoindolin-1-one

Hazard: Irritant

Formula: C₈H₆BrNO
MW: 212.0433 g/mol
MDL Number: MFCD11848599
CAS Number: 675109-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR