2-Bromo-5-hydroxy-3-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-hydroxy-3-picoline

076519-1G 076519-5G
102546-632EA 230.77 USD
102546-632 102546-634
2-Bromo-5-hydroxy-3-methylpyridine ≥95%
2-Bromo-5-hydroxy-3-methylpyridine

Hazard: Irritant

Formula: C₆H₆BrNO
MW: 188.024 g/mol
MDL Number: MFCD09839267
CAS Number: 1003711-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR