2-Amino-3-methoxybenzonitrile ≥95%

Supplier: Matrix Scientific

076518-250MG 076518-1G 076518-5G
102546-626EA 456.92 USD
102546-626 102546-628 102546-630
2-Amino-3-methoxybenzonitrile ≥95%
2-Amino-3-methoxybenzonitrile

Hazard: Irritant

Formula: C₈H₈N₂O
MDL Number: MFCD09744022
CAS Number: 148932-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR