2-(Chloromethyl)-6-methylpyridine ≥95%

Supplier: Matrix Scientific

076512-1G 076512-5G
102546-592EA 250.77 USD
102546-592 102546-594
2-(Chloromethyl)-6-methylpyridine ≥95%
2-(Chloromethyl)-6-methylpyridine

Hazard: Irritant

Formula: C₇H₈ClN
MW: 141.6000 g/mol
MDL Number: MFCD09055262
CAS Number: 3099-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR