5-Amino-1-methyl-1H-pyrazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
076510-1G
076510-5G
076510-10G
102546-580EA
280
USD
102546-580
102546-582
102546-584
5-Amino-1-methyl-1H-pyrazole-4-carboxylic acid ≥95%
5-Amino-1-methyl-1H-pyrazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₅H₇N₃O₂ MW: 141.1280 g/mol |
MDL Number:
MFCD00973833 CAS Number: 4058-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...