2-(1H-Benzo[d]imidazol-2-yl)ethanamine ≥95%

Supplier: Matrix Scientific

076508-250MG 076508-1G 076508-5G
102546-570EA 498.46 USD
102546-570 102546-572 102546-574
2-(1H-Benzo[d]imidazol-2-yl)ethanamine ≥95%
2-(1H-Benzo[d]imidazol-2-yl)ethanamine

Hazard: Irritant

Formula: C₉H₁₁N₃
MW: 161.2037 g/mol
MDL Number: MFCD00462745
CAS Number: 29518-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR