Methyl-6-bromopyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-bromopyrazine-2-carboxylate

076507-250MG 076507-1G
102546-564EA 320 USD
102546-564 102546-566
Methyl-6-bromopyrazine-2-carboxylate ≥95%
Methyl-6-bromopyrazine-2-carboxylate

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
MDL Number: MFCD14155890
CAS Number: 40155-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR