1-Boc-(2-hydroxymethyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Hydroxymethyl)piperazine, N1-BOC protected, 1-Boc-(2-Hydroxymethyl)piperazine

076506-250MG 076506-1G 076506-5G
102546-558EA 355.38 USD
102546-558 102546-560 102546-562
1-Boc-(2-hydroxymethyl)piperazine ≥95%
1-Boc-(2-hydroxymethyl)piperazine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₃
MW: 216.2800 g/mol
MDL Number: MFCD07772091
CAS Number: 205434-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR