3-Fluoro-5-nitro-1-trifluoromethylbenzene ≥95%

Supplier: Matrix Scientific

076505-1G 076505-5G
102546-552EA 92.31 USD
102546-552 102546-554
3-Fluoro-5-nitro-1-trifluoromethylbenzene ≥95%
3-Fluoro-5-nitro-1-trifluoromethylbenzene

Hazard: Irritant

Formula: C7H3F4NO2
MW: 209.0978 g/mol
MDL Number: MFCD07779168
CAS Number: 454-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR