3-(6-{[1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarbonyl]amino}-3-methylpyridin-2-yl)benzoic acid 95

Supplier: Matrix Scientific

Synonyms: Lumacaftor, 3-(6-(1-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)cyclopropanecarboxamido)-3-methylpyridin-2-yl)benzoic acid

076501-250MG 076501-1G
102546-532EA 523.08 USD
102546-532 102546-534
3-(6-{[1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarbonyl]amino}-3-methylpyridin-2-yl)benzoic acid 95
3-(6-{[1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarbonyl]amino}-3-methylpyridin-2-yl)benzoic acid

Hazard: Irritant

Formula: C₂₄H₁₈F₂N₂O₅
MW: 452.42 g/mol
MDL Number: MFCD16659051
CAS Number: 936727-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR