2-Bromo-4-fluoro-6-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

076500-1G 076500-5G 076500-10G
102546-526EA 69.23 USD
102546-526 102546-528 102546-530
2-Bromo-4-fluoro-6-methylbenzonitrile ≥95%
2-Bromo-4-fluoro-6-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₅BrFN
MW: 214.0300 g/mol
MDL Number: MFCD07782071
CAS Number: 916792-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR