Methyl-2-fluoro-4-iodonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-fluoro-4-iodonicotinate

076498-1G 076498-5G 076498-10G
102546-516EA 121.54 USD
102546-516 102546-518 102546-520
Methyl-2-fluoro-4-iodonicotinate ≥95%
Methyl-2-fluoro-4-iodonicotinate

Hazard: Irritant

Formula: C₇H₅FINO₂
MW: 281.0200 g/mol
MDL Number: MFCD04972401
CAS Number: 884494-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR