2-Bromo-6-nitroaniline ≥95%

Supplier: Matrix Scientific

076490-1G 076490-5G
102546-472EA 190.77 USD
102546-472 102546-474
2-Bromo-6-nitroaniline ≥95%
2-Bromo-6-nitroaniline

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MDL Number: MFCD09263476
CAS Number: 59255-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR