(2-Bromo-4-chlorophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-chlorophenylacetic acid

076489-1G 076489-5G
102546-466EA 190.77 USD
102546-466 102546-468
(2-Bromo-4-chlorophenyl)acetic acid ≥95%
(2-Bromo-4-chlorophenyl)acetic acid

Hazard: Irritant

Formula: C₈H₆BrClO₂
MW: 249.4900 g/mol
MDL Number: MFCD09032951
CAS Number: 52864-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR