6-Bromo-2-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2-pyridone, 2-Bromo-6-hydroxypyridine, 6-Bromo-2(1H)-pyridinone, 6-Bromo-2-pyridinol

076485-1G 076485-5G
102546-442EA 36.92 USD
102546-442 102546-444
6-Bromo-2-hydroxypyridine ≥95%
6-Bromo-2-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄BrNO
Melting Pt: 123°C
MDL Number: MFCD00234043
CAS Number: 27992-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR