2,6-Difluoro-4-methoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

076482-1G 076482-5G 076482-10G
102546-424EA 213.85 USD
102546-424 102546-426 102546-428
2,6-Difluoro-4-methoxybenzaldehyde ≥95%
2,6-Difluoro-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD04114368
CAS Number: 256417-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR