2-Phenylpyridin-4-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Amino-2-phenylpyridine
076480-250MG
076480-1G
076480-5G
102546-414EA
355.38
USD
102546-414
102546-416
102546-418
2-Phenylpyridin-4-amine ≥95%
2-Phenylpyridin-4-amine
Hazard: Irritant
Formula:
C₁₁H₁₀N₂ MW: 170.2100 g/mol |
MDL Number:
MFCD01646280 CAS Number: 21203-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...