1-Isopropylpyrrolidin-3-ylamine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Isopropyl-pyrrolidin-3-ylamine dihydrochloride, 1-(Propan-2-yl)pyrrolidin-3-amine

076479-250MG 076479-1G 076479-5G
102546-408EA 476.92 USD
102546-408 102546-410 102546-412
1-Isopropylpyrrolidin-3-ylamine dihydrochloride ≥95%
1-Isopropylpyrrolidin-3-ylamine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₈Cl₂N₂
MW: 201.1400 g/mol
MDL Number: MFCD08544449
CAS Number: 19985-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR