2,6-Quinolinediol ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxyquinolin-2(1H)-one, 2,6-Dihydroxyquinoline

076478-1G 076478-5G 076478-10G
102546-402EA 213.85 USD
102546-402 102546-404 102546-406
2,6-Quinolinediol ≥95%
2,6-Quinolinediol

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.1600 g/mol
MDL Number: MFCD00239446
CAS Number: 19315-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR