2-Iodo-4-methoxyaniline ≥95%

Supplier: Matrix Scientific

076476-1G 076476-5G 076476-10G
102546-390EA 783.08 USD
102546-390 102546-392 102546-394
2-Iodo-4-methoxyaniline ≥95%
2-Iodo-4-methoxyaniline

Hazard: Irritant

Formula: C₇H₈INO
MW: 249.05 g/mol
MDL Number: MFCD07776739
CAS Number: 191348-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR