6-Iodo-m-toluidine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-4-iodotoluene, 2-Iodo-5-methylaniline
076473-1G
076473-5G
076473-10G
102546-374EA
46.15
USD
102546-374
102546-376
102546-378
6-Iodo-m-toluidine ≥95%
6-Iodo-m-toluidine
Hazard: Irritant
Formula:
C₇H₈IN MW: 233.0496 g/mol |
MDL Number:
MFCD00833395 CAS Number: 13194-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...