2,6-Dichloro-3-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

076469-1G 076469-5G 076469-10G
102546-350EA 249.23 USD
102546-350 102546-352 102546-354
2,6-Dichloro-3-nitrobenzoic acid ≥95%
2,6-Dichloro-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₃Cl₂NO₄
MDL Number: MFCD00075422
CAS Number: 55775-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR