Ethyl adenine-9-acetate ≥95%

Supplier: Matrix Scientific

076467-1G 076467-5G
102546-342EA 640 USD
102546-342 102546-344
Ethyl adenine-9-acetate ≥95%
Ethyl adenine-9-acetate

Hazard: Irritant

Formula: C₉H₁₁N₅O₂
MW: 221.2200 g/mol
MDL Number: MFCD01321238
CAS Number: 25477-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR