4-Bromo-1,3-benzodioxole ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Bromobenzo[d][1,3]dioxole
076466-250MG
076466-1G
076466-5G
102546-336EA
456.92
USD
102546-336
102546-338
102546-340
4-Bromo-1,3-benzodioxole ≥95%
4-Bromo-1,3-benzodioxole
Hazard: Irritant
Formula:
C₇H₅BrO₂ MW: 201.0174 g/mol |
MDL Number:
MFCD02681890 CAS Number: 6698-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...