1,5-Naphthyridin-4-ol ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Hydroxy-1,5-naphthyridine
076465-250MG
076465-1G
076465-5G
102546-330EA
181.54
USD
102546-330
102546-332
102546-334
1,5-Naphthyridin-4-ol ≥95%
1,5-Naphthyridin-4-ol
Hazard: Irritant
Formula:
C₈H₆N₂O MW: 146.1460 g/mol |
MDL Number:
MFCD00234402 CAS Number: 5423-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...