Ethyl-2-(aminomethyl)isonicotinate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(aminomethyl)isonicotinate hydrochloride

076464-250MG 076464-1G 076464-5G
102546-324EA 541.54 USD
102546-324 102546-326 102546-328
Ethyl-2-(aminomethyl)isonicotinate hydrochloride ≥95%
Ethyl-2-(aminomethyl)isonicotinate hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₂
MW: 216.6700 g/mol
MDL Number: MFCD09788627
CAS Number: 1189983-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR