2-Chloro-3-nitro-6-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

076462-1G 076462-5G 076462-10G
102546-314EA 1600 USD
102546-314 102546-316 102546-318
2-Chloro-3-nitro-6-(trifluoromethyl)pyridine ≥95%
2-Chloro-3-nitro-6-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂ClF₃N₂O₂
MW: 226.5405 g/mol
MDL Number: MFCD09864674
CAS Number: 117519-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR