4,7-Dichlorothieno[2,3-d]pyridazine ≥95%

Supplier: Matrix Scientific

076459-250MG 076459-1G 076459-5G
102546-300EA 498.46 USD
102546-300 102546-302 102546-304
4,7-Dichlorothieno[2,3-d]pyridazine ≥95%
4,7-Dichlorothieno[2,3-d]pyridazine

Hazard: Irritant

Formula: C₆H₂Cl₂N₂S
MW: 205.0645 g/mol
MDL Number: MFCD09965522
CAS Number: 699-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR