7-Fluorochroman-4-one ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Fluoro-2,3-dihydro-4H-chromen-4-one

076457-1G 076457-5G
102546-290EA 146.15 USD
102546-290 102546-292
7-Fluorochroman-4-one ≥95%
7-Fluorochroman-4-one

Hazard: Irritant

Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 46…50 °C
MDL Number: MFCD06202344
CAS Number: 113209-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR