trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine ≥95%
Supplier: Matrix Scientific
076456-5G
076456-25G
102546-286EA
75.38
USD
102546-286
102664-094
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine ≥95%
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine
Hazard: Corrosive/Refrigerate
Formula:
C₈H₁₈N₂ Melting Pt: 39…44 °C |
MDL Number:
MFCD00671528 CAS Number: 87583-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...