(R)-Tetrahydrofuran-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

076455-250MG 076455-1G
102546-280EA 822.54 USD
102546-280 102546-282
(R)-Tetrahydrofuran-3-carboxylic acid ≥95%
(R)-Tetrahydrofuran-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₈O₃
MDL Number: MFCD07369940
CAS Number: 66838-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR