1H-Benzo[d]imidazol-7-amine ≥95%

Supplier: Matrix Scientific

076454-250MG 076454-1G 076454-5G
102546-274EA 481.82 USD
102546-274 102546-276 102546-278
1H-Benzo[d]imidazol-7-amine ≥95%
1H-Benzo[d]imidazol-7-amine

Hazard: Irritant

Formula: C₇H₇N₃
MW: 133.1500 g/mol
MDL Number: MFCD09054675
CAS Number: 4331-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR