(R)-tert-Butyl methyl(piperidin-3-yl)carbamate ≥95%

Supplier: Matrix Scientific

076452-250MG 076452-1G
102546-266EA 520.8 USD
102546-266 102546-268
(R)-tert-Butyl methyl(piperidin-3-yl)carbamate ≥95%
(R)-tert-Butyl methyl(piperidin-3-yl)carbamate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD09878596
CAS Number: 309962-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR