(1-Methyl-1H-benzo[d]imidazol-6-yl)methanol ≥95%
Supplier: Matrix Scientific
076450-250MG
076450-1G
076450-5G
102546-254EA
498.46
USD
102546-254
102546-256
102546-258
(1-Methyl-1H-benzo[d]imidazol-6-yl)methanol ≥95%
(1-Methyl-1H-benzo[d]imidazol-6-yl)methanol
Hazard: Irritant
Formula:
C₉H₁₀N₂O MW: 162.19 g/mol |
MDL Number:
MFCD11977685 CAS Number: 181867-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...