2,3-Dibromo-5-nitropyridine ≥95%
Supplier: Matrix Scientific
076448-1G
076448-5G
076448-10G
102546-242EA
213.85
USD
102546-242
102546-244
102546-246
2,3-Dibromo-5-nitropyridine ≥95%
2,3-Dibromo-5-nitropyridine
Hazard: Irritant
Formula:
C₅H₂Br₂N₂O₂ MW: 281.8896 g/mol |
MDL Number:
MFCD00233994 CAS Number: 15862-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...