5-Hydroxypyrimidin-4(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxypyrimidin-4(3H)-one

076447-250MG 076447-1G
102546-238EA 498.46 USD
102546-238 102546-240
5-Hydroxypyrimidin-4(1H)-one ≥95%
5-Hydroxypyrimidin-4(1H)-one

Hazard: Irritant

Formula: C4H4N2O2
MW: 112.0868 g/mol
MDL Number: MFCD11656602
CAS Number: 15837-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR