5,6-Dichloropyrimidin-4-ol ≥95%
Supplier: Matrix Scientific
Synonyms:
5,6-Dichloropyrimidin-4(3H)-one
076446-250MG
076446-1G
076446-5G
102546-232EA
427.69
USD
102546-232
102546-234
102546-236
5,6-Dichloropyrimidin-4-ol ≥95%
5,6-Dichloropyrimidin-4-ol
Hazard: Irritant
Formula:
C₄H₂Cl₂N₂O MW: 164.9775 g/mol |
MDL Number:
MFCD24393897 CAS Number: 88982-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...