4-Bromo-2-(methylthio)pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-methylsulphanyl-pyrimidine

076443-1G 076443-5G
102546-216EA 176.92 USD
102546-216 102546-218
4-Bromo-2-(methylthio)pyrimidine ≥95%
4-Bromo-2-(methylthio)pyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂S
MW: 205.07 g/mol
MDL Number: MFCD09878932
CAS Number: 959236-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR