2-Amino-4'-bromobenzophenone ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Aminophenyl)(4-bromophenyl)methanone, 2-Aminophenyl (4-bromophenyl)methanone, 2-Amino-4′-bromobenzophenone

076441-10G 076441-100G
102546-204EA 427.69 USD
102546-204 102546-206
2-Amino-4'-bromobenzophenone ≥95%
2-Amino-4'-bromobenzophenone

Hazard: Irritant

Formula: C₁₃H₁₀BrNO
MW: 276.1300 g/mol
MDL Number: MFCD02181034
CAS Number: 1140-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR