(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate HCl, Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate hydrochloride, (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylic acid Benzyl ester hydrochloride

076440-1G 076440-5G
102546-198EA 83.08 USD
102546-198 102546-200
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride ≥95%
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₀ClNO₂
MW: 281.7800 g/mol
MDL Number: MFCD04974076
CAS Number: 87269-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR