3-Fluoro-5-methylpyridine ≥95%

Supplier: Matrix Scientific

076437-1G 076437-5G
102546-180EA 38.46 USD
102546-180 102546-182
3-Fluoro-5-methylpyridine ≥95%
3-Fluoro-5-methylpyridine

Hazard: Irritant/Refrigerate

Formula: C₆H₆FN
MDL Number: MFCD03095273
CAS Number: 407-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR