2-Chloro-3-cyanobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-formylbenzonitrile

076436-1G 076436-5G
102546-176EA 364.62 USD
102546-176 102546-178
2-Chloro-3-cyanobenzaldehyde ≥95%

Hazard: Irritant

Formula: C₈H₄ClNO
MW: 165.5765 g/mol
MDL Number: MFCD08460147
CAS Number: 165187-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR