3-Methyl-1H-indazol-7-ol ≥95%

Supplier: Matrix Scientific

076434-250MG 076434-1G 076434-5G
102546-164EA 640 USD
102546-164 102546-166 102546-168
3-Methyl-1H-indazol-7-ol ≥95%
3-Methyl-1H-indazol-7-ol

Hazard: Irritant

Formula: C8H8N2O
CAS Number: 1131595-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR