5-Methoxyanthranilic acid ≥95%

Supplier: Matrix Scientific

076433-5G 076433-100G 076433-25G
102546-158EA 52.31 USD
102546-158 102546-162 102552-030
5-Methoxyanthranilic acid ≥95%
5-Methoxyanthranilic acid

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00016509
CAS Number: 6705-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR