6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

076432-250MG 076432-1G 076432-5G
102546-152EA 569.23 USD
102546-152 102546-154 102546-156
6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid ≥95%
6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
MW: 242.0300 g/mol
MDL Number: MFCD09414724
CAS Number: 1000018-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR